CAS RN 861708-66-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
methyl 2,4-dichlorothiazole-5-carboxylate, TRC
CAS: 861708-66-9 Molecular Formula: C5H3NO2SCl2 Molecular Weight (g/mol): 212.05 IUPAC Name: methyl 2,4-dichloro-1,3-thiazole-5-carboxylate SMILES: COC(=O)c1sc(Cl)nc1Cl