missing translation for 'onlineSavingsMsg'
Learn More

methyl 2,4-dichlorothiazole-5-carboxylate, TRC

Product Code. 30475601
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475601 25 mg 25mg
30472190 50 mg 50mg
30465069 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475601 Supplier Toronto Research Chemicals Supplier No. TRCM34304325MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 861708-66-9
Molecular Formula C5H3NO2SCl2
Molecular Weight (g/mol) 212.05
IUPAC Name methyl 2,4-dichloro-1,3-thiazole-5-carboxylate
SMILES COC(=O)c1sc(Cl)nc1Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H3Cl2NO2S/c1-10-4(9)2-3(6)8-5(7)11-2/h1H3
Formula Weight 210.9262
Chemical Name or Material methyl 2,4-dichlorothiazole-5-carboxylate
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.