CAS RN 86195-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(2R)-2-Isopropyl-2-(benzyloxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-2-Isopropyl-2-(benzyloxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R)-2-Isopropyl-2-(benzyloxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.