missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-Isopropyl-2-(benzyloxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt, TRC

Product Code. 30396411
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396411 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396411 Supplier Toronto Research Chemicals Supplier No. TRCI82437050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Isopropyl-2-(benzyloxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt
CAS 86195-29-1
Molecular Formula C24H32N2O9
Quantity 50 mg
Synonym (2R)-2-(1-Methylethyl)-2-(phenylmethoxy)-propanedioic Acid 1-Ethyl Ester (1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol Salt
InChI Formula InChI=1S/C15H20O5.C9H12N2O4/c1-4-19-14(18)15(11(2)3,13(16)17)20-10-12-8-6-5-7-9-12;10-8(5-12)9(13)6-1-3-7(4-2-6)11(14)15/h5-9,11H,4,10H2,1-3H3,(H,16,17);1-4,8-9,12-13H,5,10H2/t15-;8-,9-/m10/s1
SMILES CCOC(=O)[C@@](OCc1ccccc1)(C(C)C)C(=O)O.N[C@@H](CO)[C@@H](O)c2ccc(cc2)[N+](=O)[O-]
IUPAC Name (1S,2S)-2-amino-1-(4-nitrophenyl)propane-1,3-diol;(2R)-2-ethoxycarbonyl-3-methyl-2-phenylmethoxybutanoic acid
Molecular Weight (g/mol) 492.52
Formula Weight 492.2108
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.