CAS RN 861998-00-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Anamorelin Hydrochloride, TRC
CAS: 861998-00-7 Synonym: (3R)-1-(2-Methylalanyl-D-tryptophyl)-3-(phenylmethyl)-3-piperidinecarboxylic Acid Trimethylhydrazide Monohydrochloride∣RC 1291 HCl∣RC 1291 Hydrochloride
2
Anamorelin hydrochloride, MedChemExpress
MedChemExpress Anamorelin (RC-1291) hydrochloride is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.