missing translation for 'onlineSavingsMsg'
Learn More

Anamorelin hydrochloride, MedChemExpress

Product Code. 16468747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468747 5 mg 5mg
16446148 10 mg 10mg
16426536 100 mg 100mg
16422287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16468747 Supplier Medchem Express Supplier No. HY14734A
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Anamorelin (RC-1291) hydrochloride is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.

Anamorelin (RC-1291) hydrochloride is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.

Specifications

Chemical Name or Material Anamorelin hydrochloride
CAS 861998-00-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H43ClN6O3
Quantity 5 mg
Synonym RC-1291 hydrochloride ONO-7643 hydrochloride
Solubility Information DMSO : ≥ 28 mg/mL (48.01 mM)
SMILES [H][C@](CC1=CNC2=CC=CC=C21)(NC(C(C)(C)N)=O)C(N3CCC[C@@](C(N(C)N(C)C)=O)(CC4=CC=CC=C4)C3)=O.[H]Cl
Molecular Weight (g/mol) 583.16
Formula Weight 583.16
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.