CAS RN 862090-74-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chymase-IN-1, TRC
CAS: 862090-74-2 Molecular Formula: C20 H15 Cl N O4 P S Molecular Weight (g/mol): 431.829 Synonym: P-[1-(5-Chlorobenzo[b]thien-3-yl)-2-(2-naphthalenylamino)-2-oxoethyl]phosphonic Acid IUPAC Name: [1-(5-chloro-1-benzothiophen-3-yl)-2-(naphthalen-2-ylamino)-2-oxoethyl]phosphonic acid SMILES: OP(=O)(O)C(C(=O)Nc1ccc2ccccc2c1)c3csc4ccc(Cl)cc34