missing translation for 'onlineSavingsMsg'
Learn More

Chymase-IN-1, TRC

Product Code. 30365994
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365994 25 mg 25mg
30349690 2.5 mg 2.5mg
30357787 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365994 Supplier Toronto Research Chemicals Supplier No. TRCC32725525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862090-74-2
Molecular Formula C20 H15 Cl N O4 P S
Molecular Weight (g/mol) 431.829
Synonym P-[1-(5-Chlorobenzo[b]thien-3-yl)-2-(2-naphthalenylamino)-2-oxoethyl]phosphonic Acid
IUPAC Name [1-(5-chloro-1-benzothiophen-3-yl)-2-(naphthalen-2-ylamino)-2-oxoethyl]phosphonic acid
SMILES OP(=O)(O)C(C(=O)Nc1ccc2ccccc2c1)c3csc4ccc(Cl)cc34

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H15ClNO4PS/c21-14-6-8-18-16(10-14)17(11-28-18)19(27(24,25)26)20(23)22-15-7-5-12-3-1-2-4-13(12)9-15/h1-11,19H,(H,22,23)(H2,24,25,26)
Formula Weight 431.015
Chemical Name or Material Chymase-IN-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.