CAS RN 863031-24-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Azilsartan Kamedoxomil (>90%), TRC
CAS: 863031-24-7 Molecular Formula: C30H23KN4O8 Molecular Weight (g/mol): 606.62 Synonym: Azilsartan Medoxomil Monopotassium Salt IUPAC Name: potassium;(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-3-[[4-[2-(5-oxo-1-oxa-2-aza-4-azanidacyclopent-2-en-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate SMILES: [K+].CCOc1nc2cccc(C(=O)OCC3=C(C)OC(=O)O3)c2n1Cc4ccc(cc4)c5ccccc5C6=NOC(=O)[N-]6