missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan Kamedoxomil (>90%), TRC

Product Code. 30136253
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136253 25 mg 25mg
30005921 250 mg 250mg
17930925 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136253 Supplier Toronto Research Chemicals Supplier No. TRCA96560525MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863031-24-7
Molecular Formula C30H23KN4O8
Molecular Weight (g/mol) 606.62
Synonym Azilsartan Medoxomil Monopotassium Salt
IUPAC Name potassium;(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-3-[[4-[2-(5-oxo-1-oxa-2-aza-4-azanidacyclopent-2-en-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMILES [K+].CCOc1nc2cccc(C(=O)OCC3=C(C)OC(=O)O3)c2n1Cc4ccc(cc4)c5ccccc5C6=NOC(=O)[N-]6

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H24N4O8.K/c1-3-38-28-31-23-10-6-9-22(27(35)39-16-24-17(2)40-30(37)41-24)25(23)34(28)15-18-11-13-19(14-12-18)20-7-4-5-8-21(20)26-32-29(36)42-33-26;/h4-14H,3,15-16H2,1-2H3,(H,32,33,36);/q;+1/p-1
Formula Weight 606.1153
Percent Purity >95%
Chemical Name or Material Azilsartan Kamedoxomil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.