accessibility menu, dialog, popup

UserName

CH3NNNH2

CAS RN 863548-52-1

IUPAC Name: 1-(1-methyl-1H-pyrazol-5-yl)methanamine
Synonyms: 1-(2-methylpyrazol-3-yl)methanamine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: XNTFQMKXUFFUQO-UHFFFAOYSA-N
SMILES: CN1N=CC=C1CN
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)