missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%

Product Code. 15355048 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15355048 5 g 5g
15345048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15355048 Supplier Thermo Scientific Acros Supplier No. 456030050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 863548-52-1 | C5H9N3 | 111.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863548-52-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
InChI Key XNTFQMKXUFFUQO-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazol-5-yl methylamine, 1h-pyrazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-pyrazole, 2-methylpyrazol-3-yl methanamine, 5-aminomethyl-1-methylpyrazole, 1-1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazole-5-methanamine, 1-methylpyrazol-5-yl methylamine, c-2-methyl-2h-pyrazol-3-yl-methylamine
PubChem CID 7019421
IUPAC Name (2-methylpyrazol-3-yl)methanamine
SMILES CN1C(=CC=N1)CN

Specifications

Boiling Point 140.0°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methylamine
compliance-icons
Product Identifier
  • (1-Methyl-1H-pyrazol-5-yl)methylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.