accessibility menu, dialog, popup

UserName

SNOCl

CAS RN 863669-57-2

IUPAC Name: 3-(chloromethyl)-5-(thiophen-2-yl)-1,2-oxazole
Synonyms: 3-(chloromethyl)-5-(thiophen-2-yl)-1,2-oxazole
Molecular Weight (g/mol): 199.65
Molecular Formula: C8H6ClNOS
InChi Key: RKWOCISPZAHSCF-UHFFFAOYSA-N
SMILES: ClCC1=NOC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results