missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-thien-2-ylisoxazole, ≥97%, Thermo Scientific™

Product Code. 10594984
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10594984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10594984 Supplier Thermo Scientific Maybridge Supplier No. CC23124DA
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 863669-57-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H6ClNOS
MDL Number MFCD07339176
Quantity 1 g
Synonym 3-chloromethyl-5-thien-2-ylisoxazole, 3-chloromethyl-5-thiophen-2-yl-1,2-oxazole, isoxazole,3-chloromethyl-5-2-thienyl, 2-3-chloromethyl isoxazol-5-yl thiophene, 3-chloromethyl-5-thiophen-2-yl isoxazole
InChI Key RKWOCISPZAHSCF-UHFFFAOYSA-N
SMILES ClCC1=NOC(=C1)C1=CC=CS1
IUPAC Name 3-(chloromethyl)-5-thiophen-2-yl-1,2-oxazole
Molecular Weight (g/mol) 199.65
PubChem CID 7130599
Formula Weight 199.66
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-thien-2-ylisoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.