CAS RN 864068-82-6
IUPAC Name:
({imidazo[1,2-a]pyridin-6-yl}methyl)(methyl)amine
Synonyms:
{imidazo[1,2-a]pyridin-6-ylmethyl}(methyl)amine
Molecular Weight (g/mol):
161.21
Molecular Formula:
C9H11N3
InChi Key:
VTAYGJSBPFKWJX-UHFFFAOYSA-N
SMILES:
CNCC1=CN2C=CN=C2C=C1