missing translation for 'onlineSavingsMsg'
Learn More

N-(Imidazo[1,2-a]pyridin-6-ylmethyl)-N-methylamine, ≥97%, Thermo Scientific™

Product Code. 10173084
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10173084 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10173084 Supplier Thermo Scientific Maybridge Supplier No. CC33146CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 864068-82-6
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H11N3
MDL Number MFCD08271888
Quantity 250 mg
Synonym n-imidazo 1,2-a pyridin-6-ylmethyl-n-methylamine, imidazo 1,2-a pyridin-6-ylmethyl methyl amine, 1-imidazo 1,2-a pyridin-6-yl-n-methylmethanamine, 4-hydroimidazo 1,2-a pyridin-6-ylmethyl methylamine, imidazo 1,2-a pyridine-6-methanamine,n-methyl
InChI Key VTAYGJSBPFKWJX-UHFFFAOYSA-N
SMILES CNCC1=CN2C=CN=C2C=C1
IUPAC Name 1-imidazo[1,2-a]pyridin-6-yl-N-methylmethanamine
Molecular Weight (g/mol) 161.21
PubChem CID 7537497
Formula Weight 161.21
Percent Purity 95%
Chemical Name or Material N-(Imidazo[1, 2-a]pyridin-6-ylmethyl)-N-methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.