accessibility menu, dialog, popup

UserName

CH3NNN

CAS RN 864069-00-1

IUPAC Name: 2-[(4-methylpiperazin-1-yl)methyl]benzonitrile
Synonyms: 2-[(4-methylpiperazin-1-yl)methyl]benzonitrile
Molecular Weight (g/mol): 215.3
Molecular Formula: C13H17N3
InChi Key: SZENQRLSNAOEPP-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=CC=C2C#N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results