CAS RN 864071-09-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pentadecafluorooctanoic Acid-13C, TRC
CAS: 864071-09-0 Molecular Formula: 13C C7 H F15 O2 Molecular Weight (g/mol): 415.061 Synonym: Perfluorooctanoic Acid-13C (1-13C),Octanoic-1-13C acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro- (9CI) IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro(1^{13}C)octanoic acid SMILES: O[13C](=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F