missing translation for 'onlineSavingsMsg'
Learn More

Pentadecafluorooctanoic Acid-13C, TRC

Product Code. 30137258
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137258 25 mg 25mg
30136403 50 mg 50mg
30005970 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137258 Supplier Toronto Research Chemicals Supplier No. TRCP26810225MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864071-09-0
Molecular Formula 13C C7 H F15 O2
Molecular Weight (g/mol) 415.061
Synonym Perfluorooctanoic Acid-13C (1-13C), Octanoic-1-13C acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro- (9CI)
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro(1^{13}C)octanoic acid
SMILES O[13C](=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)/i1+1
Formula Weight 414.977
Percent Purity >95%
Chemical Name or Material Perfluorooctanoic Acid-13C (1-13C)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.