accessibility menu, dialog, popup

UserName

SClOCl

CAS RN 86427-02-3

IUPAC Name: 3-chlorothiophene-2-carbonyl chloride
Synonyms: 3-chlorothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H2Cl2OS
InChi Key: GCPHKTQMABHWPY-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results