missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene-2-carbonyl chloride, Thermo Scientific™

Product Code. 10663403
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10663403 1 g 1g
10039261 250 mg 250mg
10442504 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663403 Supplier Thermo Scientific Maybridge Supplier No. GK01307DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 86427-02-3
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00068137
InChI Key GCPHKTQMABHWPY-UHFFFAOYSA-N
Synonym 3-chlorothiophene-2-carbonylchloride, 3-chloro-thiophene-2-carbonyl chloride, 3-chloro-2-chlorocarbonyl thiophene, 2-thiophenecarbonyl chloride, 3-chloro-9ci, pubchem5480, timtec-bb sbb005473, buttpark 29\06-44, chloro thiophene-2-carbonyl chloride, 3-chlorothiophene-2-carboyl chloride, 3-chloro-2-thiophenecarbonyl chloride
PubChem CID 2735824
IUPAC Name 3-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=C(Cl)C=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 3-Chlorothiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.