CAS RN 864841-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tazarotene Sulfoxide, TRC
CAS: 864841-56-5 Molecular Formula: C21H21NO3S Molecular Weight (g/mol): 367.46 Synonym: Ethyl Ester 6-[2-(3,4-Dihydro-4,4-dimethyl-1-oxido-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic Acid,Ethyl 6-((4,4-Dimethyl-1-oxidothiochroman-6-yl)ethynyl)nicotinate; IUPAC Name: ethyl 6-[2-(4,4-dimethyl-1-oxo-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate SMILES: O=C(OCC)C(C=C1)=CN=C1C#CC2=CC3=C(C=C2)S(CCC3(C)C)=O