missing translation for 'onlineSavingsMsg'
Learn More

Tazarotene Sulfoxide, TRC

Product Code. 30135500
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135500 250 mg 250mg
30005349 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135500 Supplier Toronto Research Chemicals Supplier No. TRCT010090250MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864841-56-5
Molecular Formula C21H21NO3S
Molecular Weight (g/mol) 367.46
Synonym Ethyl Ester 6-[2-(3,4-Dihydro-4,4-dimethyl-1-oxido-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic Acid, Ethyl 6-((4,4-Dimethyl-1-oxidothiochroman-6-yl)ethynyl)nicotinate;
IUPAC Name ethyl 6-[2-(4,4-dimethyl-1-oxo-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
SMILES O=C(OCC)C(C=C1)=CN=C1C#CC2=CC3=C(C=C2)S(CCC3(C)C)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H21NO3S/c1-4-25-20(23)16-7-9-17(22-14-16)8-5-15-6-10-19-18(13-15)21(2,3)11-12-26(19)24/h6-7,9-10,13-14H,4,11-12H2,1-3H3
Formula Weight 367.1242
Percent Purity >95%
Chemical Name or Material Tazarotene Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.