CAS RN 864864-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CFMTI, MedChemExpress
MedChemExpress CFMTI inhibits L-glutamate-induced intracellular Ca2+ mobilization in CHO cells expressing human and rat mGluR1a, with IC50s of 2.6 and 2.3 nM, respectively.