missing translation for 'onlineSavingsMsg'
Learn More

CFMTI, MedChemExpress

Product Code. 16452576
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452576 25 mg 25mg
16425737 5 mg 5mg
16412998 10 mg 10mg
16409566 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452576 Supplier Medchem Express Supplier No. HY100402
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CFMTI inhibits L-glutamate-induced intracellular Ca2+ mobilization in CHO cells expressing human and rat mGluR1a, with IC50s of 2.6 and 2.3 nM, respectively.

CFMTI inhibits L-glutamate-induced intracellular Ca2+ mobilization in CHO cells expressing human and rat mGluR1a, with IC50s of 2.6 and 2.3 nM, respectively.

Specifications

Chemical Name or Material CFMTI
CAS 864864-17-5
Color White
Purity Grade Notes Research
Molecular Formula C19H16FN5O
Quantity 25 mg
Solubility Information DMSO : 3.7 mg/mL (10.59 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1N(C2CC2)CC3=C1C=CC(C4=C(C)N(C5=CC=CN=C5F)N=N4)=C3
Molecular Weight (g/mol) 349.36
Formula Weight 349.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.