CAS RN 864960-96-3
IUPAC Name:
3-(4-fluoro-3-methoxyphenyl)propanoic acid
Synonyms:
3-(4-fluoro-3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol):
198.19
Molecular Formula:
C10H11FO3
InChi Key:
ANPCCWWNYXSFDQ-UHFFFAOYSA-N
SMILES:
COC1=C(F)C=CC(CCC(O)=O)=C1
1
–
1
of
1
results
1
3-(4-Fluoro-3-methoxyphenyl)propionic acid, 96%, Thermo Scientific™
CAS: 864960-96-3 Molecular Formula: C10H11FO3 Molecular Weight (g/mol): 198.19 MDL Number: MFCD11655589 InChI Key: ANPCCWWNYXSFDQ-UHFFFAOYSA-N Synonym: 3-4-fluoro-3-methoxyphenyl propanoic acid,3-4-fluoro-3-methoxyphenyl propionic acid,benzenepropanoic acid, 4-fluoro-3-methoxy,3-4-fluoro-3-methoxy-phenyl-propionic acid,3-4-fluoro-3-methoxyphenyl propanoic acid im ocs PubChem CID: 21098606 IUPAC Name: 3-(4-fluoro-3-methoxyphenyl)propanoic acid SMILES: COC1=C(F)C=CC(CCC(O)=O)=C1