CAS RN 865466-24-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Narlaprevir, MedChemExpress
MedChemExpress Narlaprevir (SCH 900518) is a selective and orally bioavailable NS3 protease inhibitor with a Ki value of 6 nM and an EC90 value of 40 nM. Narlaprevir also inhibits the HCV nonstructural protein 3 serine protease. Narlaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 2.3 μM.