missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Narlaprevir, MedChemExpress
Description
Narlaprevir (SCH 900518) is a selective and orally bioavailable NS3 protease inhibitor with a Ki value of 6 nM and an EC90 value of 40 nM. Narlaprevir also inhibits the HCV nonstructural protein 3 serine protease. Narlaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 2.3 μM.
Specifications
Specifications
| Chemical Name or Material | Narlaprevir |
| CAS | 865466-24-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H61N5O7S |
| Quantity | 100 mg |
| Synonym | SCH 900518 |
| Solubility Information | DMSO : ≥ 50 mg/mL (70.63 mM) |
| SMILES | O=S(CC1(NC(N[C@H](C(N2[C@@H]([C@@]3([H])C(C)([C@]3(C2)[H])C)C(N[C@H](C(C(NC4CC4)=O)=O)CCCC)=O)=O)C(C)(C)C)=O)CCCCC1)(C(C)(C)C)=O |
| Molecular Weight (g/mol) | 707.96 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?