accessibility menu, dialog, popup

UserName

CAS RN 86620-62-4

CH3NHNOO

CAS RN 86620-62-4

IUPAC Name: 4-(4-carboxylatophenyl)-1-methylpiperazin-1-ium
Synonyms: 4-(4-carboxylatophenyl)-1-methylpiperazin-1-ium
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: UCFZVQHKTRSZMM-UHFFFAOYSA-N
SMILES: C[NH+]1CCN(CC1)C1=CC=C(C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results