CAS RN 866460-33-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Setipiprant, MedChemExpress
MedChemExpress Setipiprant is an orally available, selective CRTH2 antagonist.