CAS RN 866924-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ GW 280264X
Potent ADAM10 and ADAM17 inhibitor
2
GW 280264X, TRC
CAS: 866924-39-2 Molecular Formula: C28H41N5O6S Molecular Weight (g/mol): 575.72 Synonym: GW 280264X,BenzylN-[(5S)-5-[[(2R,3S)-3-(Formylhydroxyamino)-2-(2-methylpropyl)-1-oxohexyl]amino]-6-oxo-6-(2-thiazolylamino)hexyl]-carbamate; IUPAC Name: benzyl N-[(5S)-5-[[(2R,3S)-3-[formyl(hydroxy)amino]-2-(2-methylpropyl)hexanoyl]amino]-6-oxo-6-(1,3-thiazol-2-ylamino)hexyl]carbamate SMILES: CCC[C@@H]([C@@H](CC(C)C)C(=O)N[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)Nc2nccs2)N(O)C=O