missing translation for 'onlineSavingsMsg'
Learn More

GW 280264X, TRC

Product Code. 30240930
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240930 2.5 mg 2.5mg
30240685 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240930 Supplier Toronto Research Chemicals Supplier No. TRCG9315452.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866924-39-2
Molecular Formula C28H41N5O6S
Molecular Weight (g/mol) 575.72
Synonym GW 280264X, BenzylN-[(5S)-5-[[(2R,3S)-3-(Formylhydroxyamino)-2-(2-methylpropyl)-1-oxohexyl]amino]-6-oxo-6-(2-thiazolylamino)hexyl]-carbamate;
IUPAC Name benzyl N-[(5S)-5-[[(2R,3S)-3-[formyl(hydroxy)amino]-2-(2-methylpropyl)hexanoyl]amino]-6-oxo-6-(1,3-thiazol-2-ylamino)hexyl]carbamate
SMILES CCC[C@@H]([C@@H](CC(C)C)C(=O)N[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)Nc2nccs2)N(O)C=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C28H41N5O6S/c1-4-10-24(33(38)19-34)22(17-20(2)3)25(35)31-23(26(36)32-27-29-15-16-40-27)13-8-9-14-30-28(37)39-18-21-11-6-5-7-12-21/h5-7,11-12,15-16,19-20,22-24,38H,4,8-10,13-14,17-18H2,1-3H3,(H,30,37)(H,31,35)(H,29,32,36)/t22-,23+,24+/m1/s1
Formula Weight 575.2778
Chemical Name or Material BenzylN-((5S)-5-(((2R,3S)-3-(Formylhydroxyamino)-2-(2-methylpropyl)-1-oxohexyl)amino)-6-oxo-6-(2-thiazolylamino)hexyl)-carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.