CAS RN 868249-02-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Histidyl-Glycyl-Glutamyl-Glycyl-Threonyl-Phenylalanyl-Threonyl-Seryl-Aspartyl-Leucyl-Seryl-Lysine xTFA Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Histidyl-Glycyl-Glutamyl-Glycyl-Threonyl-Phenylalanyl-Threonyl-Seryl-Aspartyl-Leucyl-Seryl-Lysine xTFA Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Histidyl-Glycyl-Glutamyl-Glycyl-Threonyl-Phenylalanyl-Threonyl-Seryl-Aspartyl-Leucyl-Seryl-Lysine xTFA Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.