missing translation for 'onlineSavingsMsg'
Learn More

Histidyl-Glycyl-Glutamyl-Glycyl-Threonyl-Phenylalanyl-Threonyl-Seryl-Aspartyl-Leucyl-Seryl-Lysine xTFA Salt, TRC

Product Code. 30392469
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392469 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392469 Supplier Toronto Research Chemicals Supplier No. TRCH45620025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Histidyl-Glycyl-Glutamyl-Glycyl-Threonyl-Phenylalanyl-Threonyl-Seryl-Aspartyl-Leucyl-Seryl-Lysine xTFA Salt
CAS 868249-02-9
Molecular Formula C54H83N15O21 • x(C2HF3O2)
Quantity 25 mg
Synonym His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys xTFA, HGEGTFTSDLSK xTFA, 29-(2-(2-Amino-3-(1H-imidazol-5-yl)propanamido)acetamido)-2-(4-aminobutyl)-20-benzyl-11-(carboxymethyl)-17,23-bis(1-hydroxyethyl)-5,14-bis(hydroxymethyl)-8-isobutyl-4,7,10,13,16,19,22,25,28-nonaoxo-3,6,9,12,15,18,21,24,27-nonaazadotriacontane-1,32-dioic Acid xTFA
InChI Formula InChI=1S/C43H53NO15/c1-22-26(56-37(52)32(49)30(24-14-10-8-11-15-24)44-38(53)59-39(3,4)20-45)19-43(54)35(57-36(51)25-16-12-9-13-17-25)33-41(7,34(50)31(48)29(22)40(43,5)6)27(47)18-28-42(33,21-55-28)58-23(2)46/h8-17,26-28,30-33,35,45,47-49,54H,18-21H2,1-7H3,(H,44,53)/t26-,27+,28-,30+,31-,32+,33+,35-,41-,42-,43-/m1/s1
SMILES OCC(C)(C)OC(=O)N[C@@H](c1ccccc1)[C@H](O)C(=O)O[C@@H]2C[C@@]3(O)[C@H](OC(=O)c4ccccc4)[C@@H]5[C@]6(OC(C)=O)[C@H](OC6)C[C@H](O)[C@@]5(C)C(=O)[C@H](O)C(=C2C)C3(C)C
Molecular Weight (g/mol) 1278.3211402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.