CAS RN 868755-47-9
IUPAC Name:
2-bromo-1-[4-(furan-2-yl)phenyl]ethan-1-one
Synonyms:
2-bromo-1-[4-(furan-2-yl)phenyl]ethanone
Molecular Weight (g/mol):
265.11
Molecular Formula:
C12H9BrO2
InChi Key:
SONOLDBWXMKWPY-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC=C(C=C1)C1=CC=CO1