missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(2-furyl)phenyl]ethanone, Tech., Thermo Scientific™

Product Code. 10153454
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10153454 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10153454 Supplier Thermo Scientific Maybridge Supplier No. CC34405CB
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 868755-47-9
CAS Min % 80.0
CAS Max % 100.0
Molecular Formula C12H9BrO2
Linear Formula Yes
MDL Number MFCD08435854
Quantity 250 mg
Synonym 2-bromo-1-4-2-furyl phenyl ethanone, 2-bromo-1-4-furan-2-yl phenyl ethanone, 2-bromo-1-4-2-furyl phenyl ethan-1-one, ethanone,2-bromo-1-4-2-furanyl phenyl, 2-bromo-1-4-furan-2-yl phenyl ethan-1-one
InChI Key SONOLDBWXMKWPY-UHFFFAOYSA-N
SMILES C1=COC(=C1)C2=CC=C(C=C2)C(=O)CBr
IUPAC Name 2-bromo-1-[4-(furan-2-yl)phenyl]ethanone
Molecular Weight (g/mol) 265.106
PubChem CID 18525721
Formula Weight 265.11
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-[4-(2-furyl)phenyl]ethanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.