accessibility menu, dialog, popup

UserName

BHOOH

CAS RN 870774-29-1

IUPAC Name: [3-(naphthalen-2-yl)phenyl]boronic acid
Synonyms: 3-(naphthalen-2-yl)phenylboronic acid
Molecular Weight (g/mol): 248.09
Molecular Formula: C16H13BO2
InChi Key: WLKQDOGZCIYEOM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results