accessibility menu, dialog, popup

UserName

BSOONCH3CH3HOOH

CAS RN 871329-59-8

IUPAC Name: [3-(dimethylsulfamoyl)phenyl]boronic acid
Synonyms: 3-(dimethylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 229.06
Molecular Formula: C8H12BNO4S
InChi Key: SGERXKCDWJPIOS-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results