accessibility menu, dialog, popup

UserName

BFOHHOOH

CAS RN 871329-82-7

IUPAC Name: (3-fluoro-5-hydroxyphenyl)boronic acid
Synonyms: 3-fluoro-5-hydroxyphenylboronic acid
Molecular Weight (g/mol): 155.92
Molecular Formula: C6H6BFO3
InChi Key: RMBFBZIEXCTPDB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results