accessibility menu, dialog, popup

UserName

BOH3CFFFHOOH

CAS RN 871332-97-7

IUPAC Name: [3-methoxy-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 3-methoxy-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: GKEMDZIGTOAZRK-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(=C1)C(F)(F)F)B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results