accessibility menu, dialog, popup

UserName

H3COSiCH3CH3H3COCH3

CAS RN 87219-80-5

IUPAC Name: (3,3-diethoxyprop-1-yn-1-yl)trimethylsilane
Synonyms: (3,3-diethoxyprop-1-yn-1-yl)trimethylsilane
Molecular Weight (g/mol): 200.35
Molecular Formula: C10H20O2Si
InChi Key: MDRSJACTZKYOAL-UHFFFAOYSA-N
SMILES: CCOC(OCC)C#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results