missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Product Code. 15470636
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470636 5 g 5g
15480636 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470636 Supplier Thermo Scientific Alfa Aesar Supplier No. H53397.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87219-80-5
Molecular Formula C10H20O2Si
Molecular Weight (g/mol) 200.353
MDL Number MFCD08275372
InChI Key MDRSJACTZKYOAL-UHFFFAOYSA-N
Synonym 3,3-diethoxyprop-1-yn-1-yl trimethylsilane, 1-trimethylsilyl-3,3-diethoxy-1-propyne, 3-trimethylsilyl propiolaldehyde diethyl acetal, 3-trimethylsilylpropargyl aldehyde diethyl acetal
PubChem CID 11206442
IUPAC Name 3,3-diethoxyprop-1-ynyl(trimethyl)silane
SMILES CCOC(C#C[Si](C)(C)C)OCC

Specifications

Boiling Point 170°C to 172°C
Quantity 5 g
Linear Formula (CH3)3SiC≡CCH(OCH2CH3)2
UN Number UN1993
Flash Point 40°C (104°F)
Formula Weight 200.35
Percent Purity 97%
Refractive Index 1.431
Chemical Name or Material 3-(Trimethylsilyl)propiolaldehyde diethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.