accessibility menu, dialog, popup

UserName

CAS RN 873-32-5

ClN

CAS RN 873-32-5

IUPAC Name: 2-chlorobenzonitrile
Synonyms: benzonitrile, 2-chloro-
Molecular Weight (g/mol): 137.57
Molecular Formula: C7H4ClN
InChi Key: NHWQMJMIYICNBP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C#N
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)