missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzonitrile, 99%

Product Code. 10550171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10550171 1 kg 1kg
10416910 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10550171 Supplier Thermo Scientific Acros Supplier No. 108670010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 873-32-5 | C7H4ClN | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-32-5
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001779
InChI Key NHWQMJMIYICNBP-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech
PubChem CID 13391
IUPAC Name 2-chlorobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)Cl

Specifications

Melting Point 43°C to 46°C
CAS Min % 98.5
Color White to Yellow
CAS Max % 100.0
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4CN
Beilstein 09, 336
Boiling Point 232°C
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol and ether
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzonitrile, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.