accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CHNON

CAS RN 874290-95-6

IUPAC Name: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine-1-carboxamide
Synonyms: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine-1-carboxamide
Molecular Weight (g/mol): 316.21
Molecular Formula: C17H25BN2O3
InChi Key: YYWNYUFLSXVMRD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(NC(=O)N2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results