accessibility menu, dialog, popup

UserName

HNNH2FFHCl

CAS RN 875664-54-3

IUPAC Name: (3,4-difluorophenyl)hydrazine hydrochloride
Synonyms: (3,4-difluorophenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 180.58
Molecular Formula: C6H7ClF2N2
InChi Key: QTEJTSFVIILHJJ-UHFFFAOYSA-N
SMILES: Cl.NNC1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results