accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNHOH2N

CAS RN 877134-77-5

IUPAC Name: [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylurea
Molecular Weight (g/mol): 262.12
Molecular Formula: C13H19BN2O3
InChi Key: BVWCMMVUJQHJJE-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(NC(N)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results