accessibility menu, dialog, popup

UserName

CAS RN 88-65-3

HOOBr

CAS RN 88-65-3

IUPAC Name: 2-bromobenzoic acid
Synonyms: O-bromobenzoic acid
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: XRXMNWGCKISMOH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)