accessibility menu, dialog, popup

UserName

CH3ONHFF

CAS RN 881763-65-1

IUPAC Name: N-(3,5-difluorophenyl)-4-methylbenzamide
Synonyms: N-(3,5-difluorophenyl)-4-methylbenzamide
Molecular Weight (g/mol): 247.25
Molecular Formula: C14H11F2NO
InChi Key: YXZUNGODIJRLKN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NC1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results