accessibility menu, dialog, popup

UserName

CAS RN 88196-70-7

IUPAC Name: 1-(3-methoxyphenyl)ethan-1-amine
Synonyms: 1-(3-methoxyphenyl)ethanamine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: CJWGCBRQAHCVHW-UHFFFAOYNA-N
SMILES: COC1=CC=CC(=C1)C(C)N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results